Learn More
Thermo Scientific™ Piko PCR Plate, 96-well, white
Speed up PCR and qPCR applications with Piko 24-well PCR Plates, with ultra-thin wall (UTW) technology to dramatically reduce the thickness of the well walls.
$150.10 - $522.50
Products 2
Catalog Number Mfr. No. Color No. of Wells Price Quantity & Availability  
Catalog Number Mfr. No. Color No. of Wells Price Quantity & Availability  
SPL0961
Accessories View Documents
Thermo Scientific™
SPL0961
White 96 Case of 200 for $522.50
Add to cart
SPL096150
View Documents
Thermo Scientific™
SPL096150
White Case for $150.10
Add to cart
Description
Description
Piko PCR Plates are a family of PCR plates which have the footprint of a microscope slide. We have not only shrunk the size of the plate, but dramatically reduced the thickness of the well walls utilizing our patented ultra-thin wall (UTW) technology.
Highlights
  • Ultra Thin Wall (UTW) for fast PCR/qPCR applications
  • Low profile
  • Designed for use with the Piko and PikoReal 24-well thermal cyclers
  • Plates can be snapped into the plate frame to create a standard 96-well plate
  • Compatible with standard multi-channel pipettes and liquid-handling platforms
  • Well spacing and footprint conform to industry (ANSI) dimensions