Thermo Scientific™ Piko™ Thermal Cycler

Complete a PCR protocol in less than 15 minutes with the Thermo Scientific™ Piko™ Thermal Cycler, which is double the speed and half the size of its contemporaries.

Manufacturer: Thermo Scientific™ TCP0024

 View more versions of this product

Catalog No. 14-230-249

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Width (Metric) 16cm
    Block Configurations 24-well (well volume 0.225mL, sample volume max. 50µL)
    Max. Ramp Rate >5 heating and >4.5 cooling°C
    Thermal Uniformity ±0.3°C
    Thermal Accuracy ±0.2°C
    Thermal Range 4 to 99.9°C
    User Interface Semi-graphical and list mode programming
    Heated Lid Fully automated and motorized function
    No. of Programs 3861
    Depth (Metric) 17cm
    Description Piko Thermal Cycler, 24-well
    For Use With (Application) PCR, High Performance PCR
    Height (Metric) 23cm
    Weight (Metric) 4kg
    Voltage Input Power: 100 to 240V

    Complete a PCR protocol in less than 15 minutes with the Thermo Scientific™ Piko™ Thermal Cycler, which meets the highest criteria in thermal performance at twice the speed and half the size of other thermal cyclers. This is achieved using novel technical advances that allow significant reduction in PCR run times and overall size of the instrument. The Piko Thermal Cycler is an ideal solution for both convential and fast PCR applications.

    Piko Thermal Cyclers are available with two different block configurations: 24-well and 96-well. 24-well cyclers accept all standard low profile single tubes, 8-tube strips and 24-well Piko PCR Plates. The 96-well instrument utilizes 96-well Piko PCR Plates.

    • Superior thermal performance: Consistent results from well-to-well because of extremely short setting times and high temperature uniformity across the block

    • Time savings: PCR in as little as 15 minutes enabled by fast ramp rates and quick settling times

    • Reagent savings: Repeatable yields from as little as 2µL when using UTW vessels and heat sealers

    • Space savings: One of the smallest footprints available, fits into a tiny bench space

    • Energy savings: Short protocols and low wattage requires only one-fourth of the power consumption of typical thermal cyclers

    • Repeatable sealing prevents sample evaporation: Automatic heated lid provides consistent and tight sealing from run-to-run

    Recommended for:

    PCR, High Performance PCR

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit