Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Renco Pilgrim™ Powder Free White Medical Grade Exam Nitrile Glove

Pilgrim™ Nitrile Powder-Free Exam Glove incorporates our advanced proprietary formula provides excellent sensitivity, superior strength and unrivaled comfort for extended wear. Medical grade. Made in the USA.

Specifications

Color White
Grade Medical
Material Nitrile
Quantity 100/Pk., 10Pk./Cs.
Thickness (English) 3 mil
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
12-888-440A
View Documents
Renco
R12P333XS
X-Small Case of 1000 for $526.57
N/A
Due to product restrictions, please to purchase or view availability for this product.
12-888-440B
View Documents
Renco
R12P333S
Small Case of 1000 for $526.57
N/A
Due to product restrictions, please to purchase or view availability for this product.
12-888-440C
View Documents
Renco
R12P333M
Medium Case of 1000 for $526.57
N/A
Due to product restrictions, please to purchase or view availability for this product.
12-888-440D
View Documents
Renco
R12P333L
Large Case of 1000 for $526.57
N/A
Due to product restrictions, please to purchase or view availability for this product.
12-888-440E
View Documents
Renco
R12P333XL
X-Large Case of 1000 for $526.57
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

  • Advanced proprietary low modulus formulation provides premium strength, durability, and sensitivity
  • Superior stretch and comfort for extended wear
  • Polymer coated for ease of donning
  • Excellent dry & wet grip
  • Natural Rubber Latex-Free
  • Powder-Free
  • Uniform 3 mil thickness
  • Medical Grade
  • 9.5 in. Length
  • White
  • Made in the USA
Specifications

Specifications

White
Nitrile
3 mil
Medical
100/Pk., 10Pk./Cs.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit