Learn More
PILR-alpha Rabbit anti-Human, Alexa Fluor 700, Clone: 2175B, R&D Systems™
Rabbit Monoclonal Antibody
Manufacturer:  R&D Systems FAB64842N100UG
Catalog No. FAB6484N100
This item is not returnable. View return policy
Description
Description
PILR-alpha Monoclonal specifically detects PILR-alpha in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
PILR-alpha
2175B
Flow Cytometry 0.25-1 ug/10^6 cells
Q9UKJ1
Cell surface receptor FDF03, FDF03, inhibitory receptor PILRalpha, Inhibitory receptor PILR-alpha, paired immunoglobin-like receptor alpha, paired immunoglobin-like type 2 receptor alpha, paired immunoglobulin-like receptor alpha, paired immunoglobulin-like type 2 receptor alpha, PILRA, PILRalpha
Mouse myeloma cell line NS0-derived recombinant human PILR-alpha with a C-terminal 6-His tag Gln20-Thr196 Accession # Q9UKJ1
Protein A or G purified from cell culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human PILR-alpha in direct ELISAs.
Flow Cytometry
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Supernatant
Rabbit
IgG
100 ug
Primary
29992
Human