Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PILR-alpha Rabbit anti-Human, Clone: 2175B, R&D Systems™

Rabbit Monoclonal Antibody

$116.82 - $331.00

Specifications

Antigen PILR-alpha
Clone 2175B
Dilution Flow Cytometry 0.25 ug/10^6 cells, Neutralization 1-5 ug/mL
Applications Flow Cytometry, Neutralization
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB64842100 View Documents R&D Systems
MAB64842100
100 ug Each for $331.00
Add to cart
 
MAB64842SP View Documents R&D Systems
MAB64842SP
25 ug Each for $116.82
Add to cart
 
Description

Description

PILR-alpha Monoclonal antibody specifically detects PILR-alpha in Human samples. It is validated for Flow Cytometry, Neutralization.
Specifications

Specifications

PILR-alpha
Flow Cytometry 0.25 ug/10^6 cells, Neutralization 1-5 ug/mL
Unconjugated
Rabbit
Human
Q9UKJ1
29992
IgG
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human PILR-alpha in direct ELISAs.
2175B
Flow Cytometry, Neutralization
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Cell surface receptor FDF03, FDF03, inhibitory receptor PILRalpha, Inhibitory receptor PILR-alpha, paired immunoglobin-like receptor alpha, paired immunoglobin-like type 2 receptor alpha, paired immunoglobulin-like receptor alpha, paired immunoglobulin-like type 2 receptor alpha, PILRA, PILRalpha
Mouse myeloma cell line NS0-derived recombinant human PILR-alpha with a C-terminal 6-His tag Gln20-Thr196 Accession # Q9UKJ1
Monoclonal
Protein A or G purified from cell culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.

RUO

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit