Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pim2 Recombinant Mouse Monoclonal Antibody, AbFlex™, Active Motif™
Mouse Recombinant Monoclonal Antibody
Supplier: Active Motif 91239
Description
PIM proteins (PIM-1, PIM-2 and PIM-3) are oncogene-encoded serine/threonine kinases. PIM-2 is highly homologous to Pim-1 with similar oncogenic functions. PIM-2 is most abundantly expressed in hematopoietic tissues, spleen, thymus, and peripheral blood leukocytes, as well as in testis, small intestine, and colon. PIM-2 overexpression promotes resistance to a host of apoptotic stimuli; its expression is negatively regulated by growth factor depletion. Increased levels of PIM-2 has also been observed in certain cancers.Specifications
Pim2 | |
Monoclonal | |
Unconjugated | |
PIM2 | |
DXCch3; PIM2; Pim-2; pim-2 oncogene; Pim-2 proto-oncogene, serine/threonine kinase; Pim-2h; proto-oncogene Pim-2 (serine threonine kinase); proviral integration site 2; serine/threonine protein kinase pim-2; Serine/threonine-protein kinase pim-2 | |
Mouse | |
Protein A | |
RUO | |
11040 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
140mM HEPES, 32mM sodium acetate with 30% glycerol, 70mM NaCl and 0.035% sodium azide; pH 7.5 | |
Q9P1W9 | |
PIM2 | |
This antibody was raised against a recombinant protein corresponding to human PIM2 (pim-2 oncogene). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction