Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIM2 Mouse anti-Human, Clone: OTI5D5, liquid, TrueMAB™
SDP

Mouse Monoclonal Antibody

Supplier:  OriGene TA501063

Encompass_Preferred

Catalog No. 50-167-2206


Add to Cart

Description

Description

PIM proteins (PIM-1, PIM-2 and PIM-3) are oncogene-encoded serine/threonine kinases. PIM-2 is highly homologous to Pim-1 with similar oncogenic functions. PIM-2 is most abundantly expressed in hematopoietic tissues, spleen, thymus, and peripheral blood leukocytes, as well as in testis, small intestine, and colon. PIM-2 overexpression promotes resistance to a host of apoptotic stimuli; its expression is negatively regulated by growth factor depletion. Increased levels of PIM-2 has also been observed in certain cancers.
Specifications

Specifications

PIM2
Monoclonal
0.54 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9P1W9
PIM2
Full length human recombit protein of human PIM2 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG2a
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
OTI5D5
Unconjugated
PIM2
Pim-2 proto-oncogene, serine/threonine kinase
Mouse
Affinity Chromatography
RUO
11040
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit