Learn More
Pim2 Recombinant Mouse Monoclonal Antibody, AbFlex™, Active Motif™

Mouse Recombinant Monoclonal Antibody
$96.96 - $442.38
Specifications
Antigen | Pim2 |
---|---|
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3764
![]() ![]() |
Active Motif
91240 |
10 μg |
Each for $96.96
|
|
|||||
50-199-3763
![]() ![]() |
Active Motif
91239 |
100 μg |
Each for $442.38
|
|
|||||
Description
PIM proteins (PIM-1, PIM-2 and PIM-3) are oncogene-encoded serine/threonine kinases. PIM-2 is highly homologous to Pim-1 with similar oncogenic functions. PIM-2 is most abundantly expressed in hematopoietic tissues, spleen, thymus, and peripheral blood leukocytes, as well as in testis, small intestine, and colon. PIM-2 overexpression promotes resistance to a host of apoptotic stimuli; its expression is negatively regulated by growth factor depletion. Increased levels of PIM-2 has also been observed in certain cancers.Specifications
Pim2 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q9P1W9 | |
11040 | |
This antibody was raised against a recombinant protein corresponding to human PIM2 (pim-2 oncogene). | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
PIM2 |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
140mM HEPES, 32mM sodium acetate with 30% glycerol, 70mM NaCl and 0.035% sodium azide; pH 7.5 | |
DXCch3; PIM2; Pim-2; pim-2 oncogene; Pim-2 proto-oncogene, serine/threonine kinase; Pim-2h; proto-oncogene Pim-2 (serine threonine kinase); proviral integration site 2; serine/threonine protein kinase pim-2; Serine/threonine-protein kinase pim-2 | |
PIM2 | |
IgG2a | |
Protein A | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.