Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pimelic Acid, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
111-16-0
C7H12O4
160.17
MFCD00004425
WLJVNTCWHIRURA-UHFFFAOYSA-N
pimelic acid,1,5-pentanedicarboxylic acid,pileric acid,heptandioic acid,heptane-1,7-dioic acid,pimelate,6-carboxyhexanoate,6-carboxyhexanoic acid,1,7-heptanedioic acid,unii-bzq96wx25f
385
CHEBI:30531
heptanedioic acid
C(CCC(=O)O)CCC(=O)O
111-16-0
C7H12O4
160.17
MFCD00004425
WLJVNTCWHIRURA-UHFFFAOYSA-N
pimelic acid,1,5-pentanedicarboxylic acid,pileric acid,heptandioic acid,heptane-1,7-dioic acid,pimelate,6-carboxyhexanoate,6-carboxyhexanoic acid,1,7-heptanedioic acid,unii-bzq96wx25f
385
CHEBI:30531
heptanedioic acid
C(CCC(=O)O)CCC(=O)O
Melting Point 103.0°C to 106.0°C
Infrared Spectrum Authentic
Quantity 100g
Color White to Beige
Boiling Point 212.0°C (10.0 mmHg)
Packaging Glass bottle
Assay Percent Range 98%
Linear Formula HO2C(CH2)5CO2H
Beilstein 02, IV, 2003
Merck Index 15, 7543
Solubility Information Solubility in water: 50g/L (20°C). Other solubilities: freely soluble in alcohol and ether,practically insoluble in cold benzene
Formula Weight 160.17
Physical Form Powder
Percent Purity 98%
Chemical Name or Material Pimelic acid
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Wash face, hands and any exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning