Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pin1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS3541R

Description
PIN1 (Peptidylprolyl Cis/Trans Isomerase, NIMA-Interacting 1) is a Protein Coding gene. Diseases associated with PIN1 include Meibomian Cyst and Viral Exanthem. Among its related pathways are Transport of the SLBP independent Mature mRNA and Cytokine Signaling in Immune system. Gene Ontology (GO) annotations related to this gene include peptidyl-prolyl cis-trans isomerase activity and mitogen-activated protein kinase binding.Specifications
Pin1 | |
Polyclonal | |
Unconjugated | |
Pin1 | |
0610025L01Rik; BOS_45; D9Bwg1161e; dod; EC 5.2.1.8; NIMA-interacting; peptidylprolyl cis/trans isomerase, NIMA-interacting 1; peptidylprolyl cis/trans isomerase, NIMA-interacting 1 a; peptidylprolyl cis/trans isomerase, NIMA-interacting 1 S homeolog; Peptidyl-prolyl cis-trans isomerase NIMA-interacting 1; Peptidyl-prolyl cis-trans isomerase Pin1; Pin1; pin1.S; pin1a; pin1-a; pin1-b; PPIase Pin1; prolyl isomerase; prolyl isomerase Pin1; protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting 1; protein (peptidylprolyl cis/trans isomerase) NIMA-interacting 1; Rotamase Pin1; UBL5; XELAEV_18021616mg | |
Rabbit | |
Protein A | |
RUO | |
5300 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q13526 | |
Pin1 | |
KLH conjugated synthetic peptide derived from human Pin1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction