Learn More
Aviva Systems Biology Corp PIN4 Antibody - N-terminal region (ARP56687_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP56687_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PIN4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PIN4. Alternate names: EPVH, MGC138486, PAR14, PAR17ApplicationWB. Species reactivity: Human, Rat
Catalog No. 50-014-61951