Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIN4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | PIN4 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
PIN4 Polyclonal specifically detects PIN4 in Human samples. It is validated for Western Blot.Specifications
PIN4 | |
Polyclonal | |
Rabbit | |
Q9Y237-2 | |
5303 | |
IgG | |
16 kDa |
Western Blot | |
Unconjugated | |
RUO | |
4 (parvulin), MGC138486, Parvulin-17, protein (peptidylprolyl cis/trans isomerase) NIMA-interacting, 4 (parvulin) | |
PIN4 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title