Learn More
PIN4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 157891AP
Catalog No. 50-173-0479
Add to cart
Description
Description
This gene encodes a member of the parvulin subfamily of the peptidyl-prolyl cis/trans isomerase protein family. The encoded protein catalyzes the isomerization of peptidylprolyl bonds, and may play a role in the cell cycle, chromatin remodeling.
Specifications
Specifications
PIN4
0.16 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9CWW6, Q9Y237
EPVH, hEPVH, hPar14, hPar17, PAR14, PAR17, Parvulin 14, Parvulin 17, PIN4, PPIase Pin4, Rotamase Pin4
Rabbit
IgG
150 μL
-20°C
Primary
5303, 684441, 69713
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PIN4
Liquid
PIN4
PIN4 Fusion Protein Ag8489
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat