Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(+)-β-Pinene, ≥98.5% (sum of enantiomers, GC), MilliporeSigma™ Supelco™
Supplier: MilliporeSigma Supelco 806075ML
Description
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal CareSpecifications
19902-08-0 | |
Analytical Standard | |
MFCD00151105 | |
136.23 | |
Analytical Standard | |
164°C to 165°C | |
Neat |
5 mL | |
C10H16 | |
(1 R,5 R)-2(10)-Pinene; (1 R,5 R)-6,6-Dimethyl-2-methylenebicyclo[3.1.1 ]heptane | |
≥98.5% (sum of enantiomers, GC) | |
0.872 g/mL (at 25°C) | |
36°C |
Safety and Handling
ShelfLife : Limited shelf life, expiry date on the label
Recommended Storage : 2°C to 8°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction