Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pinin Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | Pinin |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP159133
|
Novus Biologicals
NBP159133 |
100 μL |
Each of 1 for $436.00
|
|
Description
Pinin Polyclonal specifically detects Pinin in Human samples. It is validated for Western Blot.Specifications
Pinin | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
140 kDa nuclear and cell adhesion-related phosphoprotein, Desmosome-associated protein, Domain-rich serine protein, DRS protein, DRSP, DRSSR-like protein, Melanoma metastasis clone A protein, MEMA, neutrophil protein, Nuclear protein SDK3, pinin, pinin, desmosome associated protein, SDK3 | |
PNN | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
Q9H307 | |
5411 | |
Synthetic peptides corresponding to PNN(pinin, desmosome associated protein) The peptide sequence was selected from the middle region of PNN. Peptide sequence RRSVDRKRRDTSGLERSHKSSKGGSSRDTKGSKDKNSRSDRKRSISESSR. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title