Learn More
Pintumomab Recombinant Mouse Monoclonal Antibody, Invitrogen™
Human Recombinant Monoclonal Antibody
Supplier: Thermo Scientific MA541874
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Pintumomab Recombinant Monoclonal antibody specifically detects Pintumomab in Chemical samples. It is validated for ELISA, Western Blot
Specifications
| Pintumomab | |
| Recombinant Monoclonal | |
| Unconjugated | |
| DPEP3 | |
| Human DPEP3 | |
| 100 μg | |
| Primary | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Human | |
| Protein A | |
| RUO | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.