Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PINX1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB100890
Description
PINX1 Polyclonal specifically detects PINX1 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
PINX1 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
FLJ20565, hepatocellular carcinoma-related putative tumor suppressor, Liver-related putative tumor suppressor, LPTLPinX1, LPTS67-11-3 protein, MGC8850, PIN2 interacting protein 1, PIN2/TERF1 interacting, telomerase inhibitor 1, PIN2/TERF1-interacting telomerase inhibitor 1, PIN2-interacting protein 1, Pin2-interacting protein X1, Protein 67-11-3, TRF1-interacting protein 1 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Expected from sequence similarity: Mouse, Rat, Pig, Dog, Cow. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:4000 | |
NP_060354.1 | |
PINX1 | |
Peptide with sequence SMLAERRRKQKWAV-C corresponding to N-Terminus according to NP_060354.4. | |
0.1 mg | |
Cancer | |
54984 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction