Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pioglitazone hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Pioglitazone hydrochloride is a selective PPARγ agonist (EC50 = 0.69 μM). Thiazolidinedione (TZD) derivative and antidiabetic agent; improves insulin sensitivity. Also enhances proliferation and survival of mESCs and acts synergistically with Y-27632 (Cat. No. 1254) to improve survival of dissociated single hPSCs and increase colony formation.
Specifications
Quantity | 10 mg |
Formula Weight | 392.9 |
Percent Purity | >99% |
Chemical Name or Material | Pioglitazone hydrochloride |
Safety and Handling
Recommended Storage | Desiccate at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction