Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIP™ ActivGrip™ Nitrile Coated Gloves with MicroFinish™ Grip
Click to view available options
Size:
Small
Medium
Large
X-Large
Description
- Revolutionary surface treatment enhances grip in dry, wet, and oily conditions
- MicroFinish™ adjusts to different conditions by reacting like tiny suction cups to prevent liquid permeation
- Seamless cotton liner increases comfort and improves fit
- Strong protection against punctures and abrasions
Specifications
Coating Material | Nitrile |
Color | Green |
Exterior Finish | No Texture |
Product Type | Dip-coated Gloves |
Size | Large |
For Use With (Application) | Fishing, Machine Operation, Metal Handling, and Oily Parts Assembly |
Glove Cuff Style | Open |
Liner Material | Cotton/Polyester Blend |
Series | 56-AG566 |
Yarn Gauge | 13 |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction