Learn More
PIP™ ActivGrip™ Nitrile Coated Gloves with MicroFinish™ Grip - 10 in.
MicroFinish™ surface coating adjusts to various conditions by reacting like tiny suction cups, making the glove impermeable to liquids.
$39.79 - $147.20
Specifications
Coating Material Nitrile
Color Gray
Glove Cuff Style Open
Liner Material Cotton/Polyester Blend
Series 56-AG585
View More Specs
Products 3
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-149-618
missing translation for 'viewDocuments'
PIP™
56AG585/S
Small
Dozen of 12 for $39.79
Case of 6 DZ for $316.50
Add to cart
19-120-804B
missing translation for 'viewDocuments'
PIP™
56AG585/M
Medium
Dozen of 12 for $147.20
Case of 6 DZ for $841.50
Add to cart
19-120-804D
missing translation for 'viewDocuments'
PIP™
56AG585/XL
X-Large
Dozen of 12 for $147.20
Case of 6 DZ for $841.50
Add to cart
Description
Description
  • Revolutionary surface treatment enhances grip in dry, wet, and oily conditions
  • MicroFinish™ adjusts to different conditions by reacting like tiny suction cups, providing a high degree of liquid resistance
  • Seamless cotton liner for increased comfort and a secure fit
  • Protection from punctures and abrasions
  • Made from component materials that comply with federal regulations for food contact 21 CFR, 170-199
Specifications
Specifications
Nitrile
Open
56-AG585
Gray
Cotton/Polyester Blend