Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Ambi-dex™ Disposable Nitrile Gloves

Industrial-grade nitrile gloves feature a textured grip for superior grasping and holding traction

Manufacturer:  Protective Industrial Products, Inc. 63338/L

 View more versions of this product

Catalog No. 19-121-200C

  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Textured
Yes
Industrial
Nitrile
4.2 in.
4 mil
Powdered
9.5 in.
9.5 x 4.2 in. (24 x 10.7cm)
Ambidextrous
Yes
Large
10.7cm
21 CFR 170-199, ISO9002
Blue
24cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit