Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Ambi-dex™ Industrial-Grade Latex Gloves

Ideal for use in laboratories, production, inspection, cleaning and light duty applications.

Manufacturer:  PIP™ 62322PF/S

Catalog No. 19-121-192A


Add to cart

Description

Description

  • Fully textured provides a good grip in wet or dry environments
  • Ambidextrous for quick and easy donning
  • Powder-Free to reduce allergic reactions
  • Beaded cuff adds additional strength to cuff edge for longer glove life and prevents liquid run-off
  • Beaded Made from component materials that comply with Federal Regulations for food contact 21 CFR, 170-199

Specifications

Specifications

ISO9001, 21 CFR 170-199, FDA
Textured
Ambidextrous
Rubber, natural
3.1 in.
5 mil
White
23.6cm
9.3 x 3.1 in. (23.6 x 7.8cm)
Beaded/Rolled
use in laboratories, production, inspection, cleaning and light duty applications
Small
7.8cm
Powder-free
9.3 in.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit