Learn More
PIP™ Ambi-dex™ Premium Industrial Grade Vinyl Gloves
Ideal for food handling, pharmaceuticals, laboratories and electronics
Manufacturer:  PIP™ 64V1000/M
 View more versions of this product
Catalog No. 19-060050
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Made from a soft virgin vinyl formulation specially compounded to deliver comfort, fit, flexibility, tactile sensitivity and performance
  • Rolled cuff adds additional strength to cuff edge for longer glove life and prevents liquid run-off
  • Contains no natural rubber latex proteins
  • Translucent white color
  • Ambidextrous
  • DEHP and DOP Free
  • 5 mil
Compliance
Made from component materials that comply with Federal Regulations for food contact 21CFR, 170-199
Specifications
Specifications
Safety
Beaded/Rolled
Industrial
Vinyl
5 mil
White
24 cm
Polymer
Ambidextrous
Yes
Medium
Powdered
9.5 in.
21 CFR 170-199