PIP™ Ambi-dex™ Regular Industrial Grade Vinyl Gloves

Ideal for food handling and processing, pharmaceuticals, laboratories and electronics

Manufacturer: Protective Industrial Products, Inc. 64-V2000/S

 View all options for this product

Catalog No. 19-023-902

  Check Availability
Add to Cart


    Color Translucent white
    Glove Cuff Style Beaded, rolled
    Hand Ambidextrous
    Grade Industrial
    Length (English) 9.5 in.
    Latex Free Yes
    Length (Metric) 24cm
    Material Vinyl
    Powdered Yes
    Size Small
    Thickness 4.5mil
    Certifications 21 CFR 170-199

    • Made from a soft virgin vinyl formulation specially compounded to deliver comfort, fit, flexibility, tactile sensitivity and performance
    • Rolled cuff adds additional strength to cuff edge for longer glove life and prevents liquid run-off
    • Contains no natural rubber latex proteins

    • Translucent white color
    • Ambidextrous
    • DEHP and DOP Free
    • 4.5 mil

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit