Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Ambi-dex™ Nitrile Disposable Powder Free Gloves

Manufacturer:  Protective Industrial Products, Inc. 63338PF/S

 View more versions of this product

Catalog No. 19-146-530



  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Textured
Yes
Ambidextrous
Yes
Small
3.4 in.
8mil
Powder-free
9.8 in.
9.8 x 3.4 in. (25 x 8.63cm)
Beaded/Rolled
Industrial
Nitrile
14
8.63cm
FDA, ISO9002
Blue
25cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit