Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ CleanTeam™ ESD Seamless-Knit Nylon Inspection Gloves

Used in circuit board manufacturing, munitions, electronics assembly and aerospace.

Manufacturer:  PIP™ 406411/S

 View more versions of this product

Catalog No. 19-121-207


Add to cart

Description

Description

  • Electrostatic Electrostatic Dissipative (ESD) fiber yarns blended with low lint nylon which reduces static build-up on the glove surface for improved performance in electronics assembly
  • Seamless knit gloves provide great dexterity
  • PVC dots provide enhanced grip and extra cut and abrasion resistance

Specifications

Specifications

ASTM D 257.2007
Dot Grip
Nylon and Carbon Fiber, PVC blend Yarn
No
Yes
Small
9.2cm
Orange
21cm
PVC
Launderable
8.3 x 3.6 in. (21 x 9.2cm)
circuit board manufacturing, munitions, electronics assembly and aerospace
Nylon, Carbon Fiber, PVC (Polyvinyl Chloride)
3.6 in.
Gray, White
8.3 in.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit