Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ ArmorFlex™ Nitrile-Coated Gloves

Lightweight and comfortable protection

$26.00

Specifications

Coated Area Palm
Color Green
Material Nitrile
For Use With (Application) general handling, assembly, fabrication, parts handling and inspection
Glove Cuff Style Knit
View More Specs

Products 5
Catalog Number Mfr. No. Length (English) Size Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Size Width (English) Price Quantity & Availability  
19-120-188B
View Documents
PIP™
563180/M
10.2 in. Medium 4.3 in.
Case of 72 for $26.00
Add to cart
 
19-120-188D
View Documents
PIP™
563180/XL
11 in. X-Large 5 in.
Case of 72 for $26.00
Add to cart
 
19-120-188A
View Documents
PIP™
563180/S
9.9 in. Small 3.9 in.
Case of 72 for $26.00
Add to cart
 
19-120-188C
View Documents
PIP™
563180/L
10.9 in. Large 4.5 in.
Case of 72 for $26.00
Add to cart
 
19-100-504
View Documents
PIP™
563182/XL
11.8 in. X-Large 4.9 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • Feature low coating-deposit weight and high abrasion resistance
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Palm coat provides flexibility and ventilation
  • Treated to reduce germs and odors
  • Chemical and cut resistant
  • Color: Green

Specifications

Specifications

Palm
Nitrile
Knit
Green
general handling, assembly, fabrication, parts handling and inspection
Cotton
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit