Learn More
PIP™ ArmorGrip Heavy-Weight Gloves
Protect against cuts, punctures, snags and abrasions
Manufacturer:  PIP™ 563145/L
 View more versions of this product
Catalog No. 19-816-808
Add to cart
Description
Description
Feature a unique combination of low coating deposit weight and high abrasion resistance. Provide long wear of heavier, bulkier nitrile-coated gloves without sacrificing flexibility and tactile sensitivity.
  • 7.6cm (3 in.) plasticized safety cuff to protect wrist and allow for fast removal
  • Cotton terry-cloth palm coated with rough-textured blue nitrile
  • ActiFresh™ treated to reduce germs and odors
  • Superior cut and abrasion resistance
For handling roofing materials, bricks and concrete blocks
Specifications
Specifications
Palm
Use commercial Detergents or laundry soap, wash in warm to medium hot Water temperature no more than 30°C for a Max. of 10 min. rinse in cold Water, tumble dry at a 60°C
Rough
use when handling roofing materials, bricks and concrete block
Cotton
5.2 in.
3mm
10.8 in.
Level 2
10.8 x 5.2 in. (27.5 x 13.3cm)
Plasticized safety
Nitrile
Large
13.3cm
Blue
27.5cm