Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIP ArmorGrip Nitrile-Coated Gloves
Heavyweight, rough-grip protection
$80.00
Specifications
Cleaning Procedure | Use commercial detergents or laundry soap, wash in warm to medium hot water temperature no more than 30°C for a Max. of 10 min. rinse in cold water, tumble dry at a 60°C |
---|---|
Color | Blue |
Material | Nitrile |
Size | Large |
For Use With (Application) | use when handling roofing materials, bricks and concrete block |
Catalog Number | Mfr. No. | Coated Area | Length (English) | Glove Cuff Style | Liner Material | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Coated Area | Length (English) | Glove Cuff Style | Liner Material | Price | Quantity & Availability | ||
19-816-809
|
PIP
563147/L |
Full | 10.8 in. | Plasticized safety | Cotton Jersey |
Dozen of 12 for $80.00
|
|
Description
- Provide the wear of heavier, bulkier nitrile-coated gloves without sacrificing flexibility and tactile sensitivity
- Feature a unique combination of low coating-deposit weight and high abrasion resistance
- Provide protection from physical hazards such as cuts, punctures and snag
- Knit wrist helps prevent dirt and debris from entering the glove
- Terry-cloth palm coated with rough-textured blue nitrile
- Treated to reduce germs and odors
Specifications
Use commercial detergents or laundry soap, wash in warm to medium hot water temperature no more than 30°C for a Max. of 10 min. rinse in cold water, tumble dry at a 60°C | |
Nitrile | |
use when handling roofing materials, bricks and concrete block | |
5.2 in. |
Blue | |
Large | |
3 mm | |
13.3 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title