Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ ArmorLite™ Textured Nitrile Coated Cotton Gloves

Gloves feature a thin nitrile coating with a textured finish for enhanced grip capabilities, while ActiFresh™ treatment reduces germs and odors.

$107.70

Specifications

Coating Material Nitrile
Color Blue, Natural
For Use With (Application) General Handling, Assembly, Fabrication, Inspection, and Maintenance
Glove Cuff Style Knit
Material Cotton
View More Specs

Products 3
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-142-756
View Documents
PIP™
563170/M
Medium
Dozen of 12 for $107.70
Case of 6 DZ for $615.50
Add to cart
 
19-142-757
View Documents
PIP™
563170/L
Large
Dozen of 12 for $107.70
Case of 6 DZ for $615.50
Edge
Add to cart
 
19-142-758
View Documents
PIP™
563170/XL
X-Large
Dozen of 12 for $107.70
Case of 6 DZ for $615.50
Edge
Add to cart
 
Description

Description

  • Cotton interlock liner thinly coated with blue nitrile
  • Comfortable and flexible for a high degree of dexterity
  • Textured coating provides enhanced grip capabilites
  • Effective, affordable protection from physical hazards such as cut, puncture, snags and abrasions
  • Resistant to grease and oils
  • Made in ISO 9001:2000 / ISO 14001:2000 facilities
  • Launderable
Specifications

Specifications

Nitrile
General Handling, Assembly, Fabrication, Inspection, and Maintenance
Cotton
Blue, Natural
Knit
56-3170
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit