Learn More
PIP™ ArmorLite™ Thinly Coated Nitrile-Dipped Gloves
Comfortable and flexible to allow for excellent dexterity
Manufacturer:  PIP™ 563176/M
Catalog No. 19-121-189
Add to cart
Description
Description
  • 7.6cm (3 in.) plasticized safety cuff helps to protect wrist and allows for fast removal
  • Effective protection from physical hazards such as cuts, punctures, snags and abrasions
  • Cotton interlock liner, thinly coated with blue nitrile
  • ActiFresh™ treated to reduce germs and odors
  • Textured coating provides grip
  • Resistant to grease and oils
  • Launderable
  • Size: Medium
  • Color: Blue
General handling, assembly, fabrication, parts handling, inspection, maintenance, shipping and receiving
Specifications
Specifications
Launderable
Plasticized safety
Nitrile
Medium
Textured
general handling, assembly, fabrication, parts handling, inspection, maintenance and shipping and receiving
Cotton
Blue