Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ ArmorLiteXT™ Textured Nitrile Coated Cotton Gloves - Knit Wrist

Catalog Number 19161893
Click to view available options
Size:
Small
Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Armor™ Series gloves combine low-coating deposit weight and high abrasion resistance to provide the wear of heavier, bulkier nitrile coated gloves without sacrificing flexibility and tactile sensitivity.

  • ActiFresh™ treated to reduce germs and odors
  • Gloves enhance dexterity, chemical and cut resistance
  • Palm coating provides flexibility and ventilation
  • Economic protection for handling small, oily parts
  • Knit wrist prevents dirt and debris from entering the glove
  • Manufactured in ISO 9001:2000/ISO 14001:2000 facilities
Coated Area Palm, Fingers, Knuckles
Coating Material Nitrile
Color Green
Exterior Finish Fully Textured
Hand Specific
Material Cotton
Product Type Dip-coated Gloves
Size Small
For Use With (Application) General Assembly, Fabrication, Parts Handling, and Inspection
Glove Cuff Color Natural
Glove Cuff Style Knit
Series 56-3775
Length (English) 9.6 in.
Length (Metric) 24.5 cm
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit