Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ ArmorTuff™ Nitrile Coated Jersey Gloves - Knit Wrist

Armor™ Series gloves combine low-coating deposit weight with high abrasion resistance to provide the protection of heavy-duty coated gloves, without sacrificing flexibility or tactile sensitivity.

$53.00

Specifications

Coating Material Nitrile
Color Blue, Natural
For Use With (Application) General Handling, Assembly, Fabrication, and Maintenance
Glove Cuff Style Knit
Material Cotton
View More Specs

Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-148-277
View Documents
PIP™
563151/XL
X-Large
Dozen of 12 for $53.00
Case of 6 DZ for $303.00
Add to cart
 
19-148-275
View Documents
PIP™
563151/M
Medium This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • ActiFresh™ treated to reduce germs and odors
  • Protects from physical hazards such as cuts, punctures, snags, and abrasions
  • Cotton jersey liner with blue nitrile coating
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Resistant to grease and oils
  • Launderable
Specifications

Specifications

Nitrile
General Handling, Assembly, Fabrication, and Maintenance
Cotton
Blue, Natural
Knit
56-3151
SDS
Product Certifications

WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit