Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ ArmorTuff™ Standard-Weight Nitrile-Dipped Gloves

Comfortable and flexible

$8.66 - $77.00

Specifications

Cleaning Procedure Launderable
Cut Resistance Rating Level 2
For Use With (Application) general handling, assembly, fabrication, parts handling and maintenance
Liner Material Cotton Jersey
Color Blue
View More Specs

Products 3
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
19-121-188C View Documents PIP™
563152/L
N/A Large; Coating, Palm Dozen of 12 for $8.66
Case of 6 DZ for $51.96
Add to cart
 
19-121-188D View Documents PIP™
563152/XL
N/A X-Large; Coating, Palm Dozen of 12 for $77.00
Case of 6 DZ for $434.00
Add to cart
 
19-816-814 View Documents PIP™
563154/L
N/A Plasticized cuff; Large; Coating, Full Dozen of 12 for $48.71
Add to cart
 
Description

Description

  • Blue nitrile coating on cotton jersey liner
  • Knit wrist or 7.6cm (3 in.) plasticized safety cuff
  • Provide excellent dexterity and grip
  • Resistant to grease and oils
  • Machine washable
  • Color: Blue

Ideal for general handling, assembly, fabrication, parts handling and maintenance

Specifications

Specifications

Launderable
general handling, assembly, fabrication, parts handling and maintenance
Blue
Level 2
Cotton Jersey
Nitrile
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit