Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Assurance™ 52-3672 Unsupported Neoprene Coated Latex Gloves with Raised Diamond Grip

Catalog Number 19161646A
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Double-dipped neoprene construction provides protection against a wide range of chemicals such as acids, caustics, solvents, and oils. Raised diamond pattern enhances grip capabilities in wet or oily conditions.

  • Neoprene coating provides protection from a wide range of chemicals such as acids, caustics, solvents, and oils
  • Double dipped construction combines the versatile chemical resistance properties of neoprene with the comfort of latex
  • Straight cuff adds additional length to protect the forearm from liquid run-off
  • Flock lining increases comfort and eases donning/doffing
  • Raised diamond pattern enhances grip capabilities
  • All component materials comply with Federal regulations for food contact 21 CFR, 170-199
Certifications/Compliance 21 CFR, 170-199
Coating Material Neoprene
Color Blue, Yellow
Exterior Finish Partially Textured
Material Latex
Product Type Dip-coated Gloves
Size Small
Thickness (English) 19 mil
EN388 Standard 1X11
For Use With (Application) General Maintenance, Janitorial Work, Petrochemical, and Photo Finishing
Glove Cuff Style Straight
Liner Material Flocked Cotton
Series 52-3672
Length (English) 12.6 in.
Length (Metric) 32 cm
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit