Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Assurance™ Unsupported 11mil Lightweight Nitrile Gloves, Unlined

Provides protection from chemical and physical hazards

$44.15

Specifications

Certifications/Compliance ISO9001:2000, ISO14001:2004, 21 CFR 170-199, FDA, CE
Coating Material Raised diamond
Color Green
Length (English) 13 in.
Length (Metric) 33 cm
View More Specs

Products 5
Catalog Number Mfr. No. Size Powdered Width (English) Width (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Size Powdered Width (English) Width (Metric) Price Quantity & Availability  
19-162-727
View Documents
PIP™
50N110G/M
Medium Powdered 2.8 in. 7 cm
Dozen of 12 for $44.15
Case of 12 DZ for $492.00
Add to cart
 
19-162-726
View Documents
PIP™
50N110G/S
Small Powdered 2.7 in. 6.8 cm
Dozen of 12 for $44.15
Case of 12 DZ for $492.00
Add to cart
 
19-162-729
View Documents
PIP™
50N110G/XL
X-Large Powdered 3.3 in. 8.4 cm
Dozen of 12 for $44.15
Case of 12 DZ for $492.00
Add to cart
 
19-162-730
View Documents
PIP™
50N110G/XXL
2X-Large Powder-free 3.5 in. 9 cm
Dozen of 12 for $44.15
Case of 12 DZ for $492.00
Add to cart
 
19-162-728
View Documents
PIP™
50N110G/L
Large Powdered 3.1 in. 7.8 cm
Dozen of 12 for $44.15
Case of 12 DZ for $492.00
Add to cart
 
Description

Description

  • Light weight construction and hand specific design provide real comfort with protection
  • Provides flexibility, tactile sensitivity and dexterity sometimes lacking in heavier gauge unsupported nitriles
  • Raised diamond grip pattern provides superior grip
  • Unlined to reduce potential of product contamination
  • Straight cuff
  • Chlorinated
  • Length: 13 in. (33cm)
  • Color: green

Light chemical applications, printing and meat and poultry processing

Certifications

ISO 9001:2000

Compliance

Federal regulations

Specifications

Specifications

ISO9001:2000, ISO14001:2004, 21 CFR 170-199, FDA, CE
Green
33 cm
Light chemical applications, printing, meat and poultry processing
11 mil
Raised diamond
13 in.
Nitrile
Straight
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit