PIP™ Assurance™ Unsupported 15mil Lightweight Nitrile Gloves, Flock-Lined

Provide superior resistance to physical hazards

Manufacturer: Protective Industrial Products, Inc. 50-N160B/XL

 View all options for this product

Catalog No. 19-161-851

  Check Availability
Add to Cart


    Coating Material Raised diamond
    Color Blue
    Dimensions (L x W) 13 x 3.3 in. (33 x 8.4cm)
    Glove Cuff Style Straight
    For Use With (Application) cleaning and de-greasing, graphic arts and printing cleanup, handling equipment and abrasive materials coated with Oil, Grease, Acids and Solvents, handling fillers, Solvents, dye agents, cleaning Solvents, cement and mortar mixing
    Length (English) 13 in.
    Length (Metric) 33cm
    Lining Flock
    Material Nitrile
    Size X-Large
    Thickness 15mil
    Width (English) 3.3 in.
    Width (Metric) 8.4cm
    Certifications ISO9001:2000, 21 CFR 170-199, FDA, CE

    Gloves provide resistance to cut, puncture, snag and abrasion and many chemical hazards, including solvents and oil-based chemicals.

    • Light weight construction and hand specific design provide real comfort with protection
    • Provides flexibility, tactile sensitivity and dexterity sometimes lacking in heavier gauge unsupported nitriles
    • Raised diamond grip pattern provides superior grip
    • Chlorinated
    • Straight cuff
    • Length: 13 in. (33cm)
    • Thickness: 15mil
    • Color: green or blue


    Light chemical applications, printing and meat and poultry processing.

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit