Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Assurance™ Unsupported 15mil Lightweight Nitrile Gloves, Unlined

Provides the protection from chemical and physical hazards

$42.50 - $51.50

Specifications

For Use With (Application) light chemical applications, printing, meat and poultry processing
Thickness (English) 15mil
View More Specs

Products 4
Catalog Number Mfr. No. Size Color Price Quantity & Availability  
Catalog Number Mfr. No. Size Color Price Quantity & Availability  
19-152-515
View Documents
PIP™
50N140B/XXL
2X-Large Blue
Dozen of 12 for $42.50
Case of 12 DZ for $403.00
Add to cart
 
19-152-517
View Documents
PIP™
50N140G/M
Medium Green
Dozen of 12 for $51.50
Case of 12 DZ for $580.08
Add to cart
 
19-152-519
View Documents
PIP™
50N140G/XXL
2X-Large
Dozen of 12 for $51.50
Case of 12 DZ for $581.22
Add to cart
 
19-152-518
View Documents
PIP™
50N140G/L
Large Green
Dozen of 12 for $51.50
Case of 12 DZ for $580.08
Add to cart
 
Description

Description

  • Made from component materials which comply with Federal regulations for food contact; 21CFR, 170-199
  • Light weight construction and hand specific design provide real comfort with protection
  • Provides flexibility, tactile sensitivity and dexterity sometimes lacking in heavier gauge unsupported nitriles
  • Raised diamond grip pattern provides superior grip
  • Unlined to reduce potential of product contamination
  • Straight cuff
  • Chlorinated
  • Length: 13 in. (33cm)
  • Color: blue or green
  • Light chemical applications, printing and meat and poultry processing

Specifications

Specifications

light chemical applications, printing, meat and poultry processing
15mil
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit