Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Assurance™ Unsupported 15mil Medium and Heavy-weight Nitrile Gloves, Flock-Lined

Provide protection from physical and chemical hazards such as cut, puncture, solvents and oil-based chemicals

Specifications

Certifications/Compliance ISO9001:2000, 21 CFR 170-199, FDA
Coating Material Diamond
Color Green
Length (English) 13 in.
Length (Metric) 33 cm
View More Specs

Products 3
Catalog Number Mfr. No. Size Powdered Width (English) Width (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Size Powdered Width (English) Width (Metric) Price Quantity & Availability  
19-162-733
View Documents
PIP™
50N155G/L
Large Powdered 4.8 in. 12.2 cm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-162-734
View Documents
PIP™
50N155G/XL
X-Large Powdered 5.4 in. 13.7 cm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-162-731
View Documents
PIP™
50N155G/S
Small Powder-free 4 in. 10.16 cm This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • Thickness: 15mil
  • Flock Lined
  • Super diamond grip
  • Straight cuff
  • Color: green
  • Length: 13 in. (33cm)

  • Chemical processing
  • Oil refining and petrochemicals
  • Food processing
  • Aerospace and automotive degreasing
  • Automotive assembly and painting
  • Machining operations
  • Metal fabrication graphic arts and printing cleanup
  • Furniture manufacturing and electronics

Certifications

ISO 9001:2000

Compliance

Federal Regulations

Specifications

Specifications

ISO9001:2000, 21 CFR 170-199, FDA
Green
33 cm
chemical processing, Oil refining and Petrochemicals, food processing, aerospace and automotive degreasing, automotive assembly and painting, machining operations, Metal fabrication graphic arts and printing cleanup, furniture manufacturing and
Flock
Diamond
13 in.
Nitrile
Straight
15mil
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit