PIP™ Assurance™ Unsupported 15mil Medium and Heavy-weight Nitrile Gloves, Flock-Lined

Provide protection from physical and chemical hazards such as cut, puncture, solvents and oil-based chemicals

Manufacturer: Protective Industrial Products, Inc. 50N155G/S

 View more versions of this product

Catalog No. 19-162-731

  Check Availability
Add to Cart

    Specifications

    Certifications ISO9001:2000, 21 CFR 170-199, FDA
    Dimensions (L x W) 13 x 4 in. (33 x 10.16cm)
    Glove Cuff Style Straight
    For Use With (Application) chemical processing, Oil refining and Petrochemicals, food processing, aerospace and automotive degreasing, automotive assembly and painting, machining operations, Metal fabrication graphic arts and printing cleanup, furniture manufacturing and
    Thickness 15mil
    Coating Material Diamond
    Length (English) 13 in.
    Length (Metric) 33cm
    Lining Flock
    Material Nitrile
    Powdered No
    Size Small
    Width (English) 4 in.
    Width (Metric) 10.16cm
    Color Green

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit