Learn More
PIP™ Assurance™ Unsupported 18mil Latex Gloves, Flock Lined
Flock lining provides increased comfort and ease of donning
$1.63 - $31.75
Save up to 75%
Specifications
Certifications/Compliance ISO9001:2000, 21 CFR 170-199, FDA
Length (English) 12 in.
Length (Metric) 30.5 cm
Material Cotton, Rubber Latex
Exterior Finish Honeycomb
View More Specs
Alert:
This style contains natural rubber latex which may cause allergic reactions. Safe use by or on latex-sensitive individuals has not been established.
Products 11
Catalog Number Mfr. No. Color Size Glove Cuff Style Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Color Size Glove Cuff Style Width (English) Price Quantity & Availability  
19-162-713
View Documents
PIP™
48L185B/S
Blue Small Pinked 3.5 in. Dozen of 12 for $30.80
Case of 12 DZ for $356.00
Add to cart
19-162-714
View Documents
PIP™
48L185B/M
Blue Medium Pinked 3.9 in. Dozen of 12 for $30.80
Case of 12 DZ for $356.00
Add to cart
19-162-717
View Documents
PIP™
48L185T/S
Orange Small Beaded/Rolled 3.5 in. Dozen of 12 for $29.85
Case of 12 DZ for $284.00
Add to cart
19-162-718
View Documents
PIP™
48L185T/M
Orange Medium Beaded/Rolled 3.9 in. Dozen of 12 for $29.85
Case of 12 DZ for $284.00
Add to cart
19-162-715
View Documents
PIP™
48L185B/L
Blue Large Pinked 4.3 in. Dozen of 12 for $31.75
Case of 12 DZ for $349.00
Add to cart
19-162-716
View Documents
PIP™
48L185B/XL
Blue X-Large Pinked 4.6 in. Dozen of 12 for $1.63
Save 75% Reg : $6.51
Case of 12 DZ for $19.50
Save 95% Reg : $356.00
Add to cart
19-162-720
View Documents
PIP™
48L185T/XL
Orange X-Large Beaded/Rolled 4.6 in. Dozen of 12 for $29.85
Case of 12 DZ for $284.00
Add to cart
19-152-521
View Documents
PIP™
48L185P/S
Pink Small Pinked 3.5 in. Dozen of 12 for $30.80
Case of 12 DZ for $356.00
Add to cart
19-152-522
View Documents
PIP™
48L185P/M
Pink Medium Pinked 3.9 in. Dozen of 12 for $30.80
Case of 12 DZ for $356.00
Add to cart
19-152-523
View Documents
PIP™
48L185P/L
Pink Large Pinked 4.3 in. Dozen of 12 for $30.80
Case of 12 DZ for $356.00
Add to cart
19-152-524
View Documents
PIP™
48L185P/XL
Pink X-Large Pinked 4.6 in. Dozen of 12 for $30.80
Case of 12 DZ for $356.00
Add to cart
Description
Description
  • Honeycomb grip
  • Available with pinked and rolled bead cuff
  • Thickness: 18mil
  • Length: 12 in. (30.4cm)
  • Color: pink
It is strongly advised not to wash and re-use these gloves when they are used to protect against chemical products
Janitorial, sanitation and general maintenance.
Certifications
ISO 9001:2000
Specifications
Specifications
ISO9001:2000, 21 CFR 170-199, FDA
30.5 cm
Honeycomb
Flock
18 mil
12 in.
Cotton, Rubber Latex
janitorial, sanitation and general maintenance
Powder-free