PIP™ Assurance™ Unsupported 21mil Medium and Heavy Weight Nitrile Gloves, Flock Lined

Provides resistance to physical hazards such as cut, puncture, snag and abrasion and many chemical hazards including solvents and oil-based chemicals

Manufacturer: Protective Industrial Products, Inc. 50N2420GY/XL

 View more versions of this product

Catalog No. 19-162-737

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Certifications ISO9001:2000, 21 CFR 170-199, FDA
    Dimensions (L x W) 12 x 5.2 in. (30.5 x 13.2cm)
    Glove Cuff Style Straight
    For Use With (Application) chemical processing, Oil refining and Petrochemicals, food processing, aerospace and automotive degreasing, automotive assembly and painting, machining operations, Metal fabrication, graphic arts and printing cleanup, furniture manufacturing and
    Length (English) 12 in.
    Length (Metric) 30.5cm
    Lining Flock
    Material Nitrile
    Powdered No
    Size X-Large
    Thickness 21mil
    Coating Material Raised diamond
    Width (English) 5.2 in.
    Width (Metric) 13.2cm
    Color Gold, yellow

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit