Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Assurance™ Unsupported 21mil Medium and Heavy Weight Nitrile Gloves, Flock Lined

Provides resistance to physical hazards such as cut, puncture, snag and abrasion and many chemical hazards including solvents and oil-based chemicals

Specifications

Certifications/Compliance ISO9001:2000, 21 CFR 170-199, FDA
Coating Material Raised diamond
Color Gold, Yellow
Length (English) 12 in.
Length (Metric) 30.5 cm
View More Specs

Products 2
Catalog Number Mfr. No. Size Powdered Price Quantity & Availability  
Catalog Number Mfr. No. Size Powdered Price Quantity & Availability  
19-162-736
View Documents
PIP™
50N2420GY/M
Medium Powdered This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
19-161-843
View Documents
PIP™
50N2420GY/S
Small Powdered This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

  • Thickness: 21mil
  • Flock lined
  • Raised diamond grip
  • Straight cuff
  • Color: gold over yellow
  • Length12 in. (30.48cm)
  • It is strongly advised not to wash and re-use these gloves when they are used to protect against chemical products.

    • Chemical processing
    • Oil refining and petrochemicals
    • Food processing
    • Aerospace and automotive degreasing
    • Automotive assembly and painting
    • Machining operations
    • Metal fabrication
    • Graphic arts
    • Printing cleanup
    • Furniture manufacturing
    • Electronics

    Certifications

    ISO 9001:2000

    Compliance

    Federal regulations

Specifications

Specifications

ISO9001:2000, 21 CFR 170-199, FDA
Gold, Yellow
30.5 cm
chemical processing, Oil refining and Petrochemicals, food processing, aerospace and automotive degreasing, automotive assembly and painting, machining operations, Metal fabrication, graphic arts and printing cleanup, furniture manufacturing and
Flock
Raised diamond
12 in.
Nitrile
Straight
21 mil
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit