PIP™ ATG™ MaxiChem™ Cut™ Nitrile Blend Coated Gloves

Developed for people working with chemicals while also requiring a level of cut protection

Manufacturer: Protective Industrial Products, Inc. 56633/S

 View more versions of this product

Catalog No. 19-148-207

  Check Availability
Add to Cart

    Specifications

    Palm Type Non-slip Grip
    Dimensions (L x W) 30.5 x 10.4cm (12 x 4.1 in.)
    Exterior Finish Micro-foam nitrile
    Glove Cuff Style Gauntlet
    Lining HPPE
    Size Small
    Color Green/Black

    • Chemical resistance delivered through ATG LiquiTech™ technology platform
    • Seamless knit engineered yarn
    • Ultra lightweight micro-foam nitrile coating
    • Super light seamless knit HPPE liner provides comfort, sensitivity, fit and dexterity
    • Gauntlet cuff keeps cuff close to hand to maintain fingers at tip of gloves to maximize dexterity
    • Micro-cup non-slip grip finish on palm for controlled and superior grip in oily and wet applications
    • All manufacturing ingredients comply with REACH legislation
    • Green shell, black grip and white liner

    Applications:

    General handling and handling small castings, handling of small components, light metal fabrication and parts handling.

    Certifications

    CE.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit