Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ ATG™ MaxiDry™ Hi-Performance Nitrile Gloves

Ultra thin design provides high comfort and flexibility along with maximum finger tip tactile sensitivity

$107.80 - $156.10

Specifications

Cleaning Procedure Recommend washing gloves in slightly warm water not exceeding 40°C, a mild commercial laundry soap or detergent should be used, wash in a 5 to 10 min. cycle time, rinse in cold water, tumble dry at temperature not exceeding 60°C
Color Black, Purple
Cut Resistance Rating Level 1
Length (English) 9.8 in.
Length (Metric) 25 cm
View More Specs

Products 10
Catalog Number Mfr. No. Coated Area Size Glove Cuff Style Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Coated Area Size Glove Cuff Style Width (English) Price Quantity & Availability  
19-148-457
View Documents
PIP™
56424/M
Palm Medium Continuous knit 3.3 in. Dozen of 12 for $156.10
Case of 6 DZ for $646.00
Add to cart
 
19-148-456
View Documents
PIP™
56424/S
Palm Small Continuous knit 3.2 in. Dozen of 12 for $156.10
Case of 6 DZ for $646.00
Add to cart
 
19-148-458
View Documents
PIP™
56424/L
Palm Large Continuous knit 3.7 in. Dozen of 12 for $107.80
Case of 6 DZ for $587.00
Add to cart
 
19-148-460
View Documents
PIP™
56424/XXL
Palm 2X-Large Continuous knit 3.9 in. Dozen of 12 for $156.10
Case of 6 DZ for $646.00
Add to cart
 
19-148-459
View Documents
PIP™
56424/XL
Palm X-Large Continuous knit 3.8 in. Dozen of 12 for $107.80
Case of 6 DZ for $587.00
Add to cart
 
19-148-462
View Documents
PIP™
56425/M
0.75 Dipped Medium Gauntlet 3.3 in. Dozen of 12 for $123.20
Case of 6 DZ for $678.50
Add to cart
 
19-148-461
View Documents
PIP™
56425/S
0.75 Dipped Small Gauntlet 3.2 in. Dozen of 12 for $123.00
Case of 6 DZ for $671.00
Add to cart
 
19-148-463
View Documents
PIP™
56425/L
0.75 Dipped Large Gauntlet 3.7 in. Dozen of 12 for $123.20
Case of 6 DZ for $678.50
Add to cart
 
19-148-464
View Documents
PIP™
56425/XL
0.75 Dipped X-Large Gauntlet 3.8 in. Dozen of 12 for $123.00
Case of 6 DZ for $671.00
Add to cart
 
19-148-465
View Documents
PIP™
56425/XXL
0.75 Dipped 2X-Large Gauntlet 3.9 in. Dozen of 12 for $123.00
Case of 6 DZ for $671.00
Add to cart
 
Description

Description

  • Ultra lightweight nitrile on fine gauge seamless knit nylon liner
  • GripTech™ coating offers non-slip grip
  • Continuous knit, locking cuff ensures the glove doesn't slip on the hand
  • EN certified 100% low level chemical and oil repellant – EN 374-3
  • LiquiTech™ Barrier Technology provides resistant to oils and liquids
  • Silicone free
  • Color: Purple and Black
  • Length: 10 in. (25.4cm)

Model: 56-425
3/4 dipped coated

Applications:

Precision handling in oily or wet environments; Engine, and power train, Gear box and transmission assembly, General handling, handling small castings, handling of small components, light metal fabrication and parts handling, primary, secondary and final assembly, shock absorber assembly, maintenance.

Specifications

Specifications

Recommend washing gloves in slightly warm water not exceeding 40°C, a mild commercial laundry soap or detergent should be used, wash in a 5 to 10 min. cycle time, rinse in cold water, tumble dry at temperature not exceeding 60°C
Level 1
25 cm
Non-slip
Nylon
Black, Purple
9.8 in.
Lycra™, Nitrile
precision handling in Oily or wet environments, offshore Petroleum, light Metal fabrication and part handling, engine, power train and transmission assembly, handling small castings, gears and other abrasive edged parts, light stamping in Oily c
1.3 mm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit