PIP™ Bolle' Contour Safety Eyewear
High impact 2.2mm polycarbonate lens with anti-fog/anti-scratch coated on both sides
Manufacturer: Protective Industrial Products, Inc. 253-CT-40047
 View all options for this product
Catalog No. 19-036-816
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    For Use With (Application) Assembly and production work; Automotive; Solar UV exposure; Emergency response; Rigging; Laboratory and dental work; Telecommunications
    Frame Material Nylon
    Lens Material Polycarbonate
    Lens Tint Extra Sensory Perception Tinted
    Frame Color Dark Gunmetal
    Lens Coating Anti-scratch; Anti-fog
    Ultra lightweight half frame provides extensive eye coverage.
    • High contour (8 base), dual lens piece
    • Offers 180° distortion free viewing and superior optical quality
    • 99.99% UVA and UVB light absorption
    • Optimal fit and comfort with non-slip temple tips
    • Reinforced nose bridge
    • Non-slip rubber nose pads
    • Supplied with Soft pouch and adjustable cord
    • ESP lens reduces eye fatigue and eye strain caused by blue light
    Assembly and production work, Automotive, Solar UV exposure, Emergency response, Rigging, Laboratory and dental work, Telecommunications
  • Description & Specifications
Related Products
Product Content Feedback