PIP™ Bolle's Coverall Safety Goggles
High impact 1.5mm polycarbonate lens with anti-fog/anti-scratch coating on both sides
Manufacturer: Protective Industrial Products, Inc. 254CL40101
 View more versions of this product
Catalog No. 19-036-868
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Frame Color White
    For Use With (Application) Chemical splash and dust; protection; Engineering; Process work; Cleaning Services
    Frame Material TPR
    Lens Material Polycarbonate
    Lens Tint Clear
  • Description & Specifications
Related Products
Product Content Feedback