PIP™ Bolle's Safety Atom Protective Eyewear
High impact 2.2mm polycarbonate lens with anti-fog/anti-scratch coating on both sides
Manufacturer: Protective Industrial Products, Inc. 254AT40092
 View more versions of this product
Catalog No. 19-036-861
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Frame Color Blue
    Lens Coating Anti-scratch; Anti-fog
    For Use With (Application) Assembly and production work; Process work; Emergency response; Drilling; Painting
    Frame Material Polyvinyl Chloride (PVC)
    Includes Adjustable strap
    Lens Material Polycarbonate
    Lens Tint Clear
  • Description & Specifications
Related Products
Product Content Feedback