PIP™ Bolle's Safety Override Protective Eyewear
Fits over prescription eyewear to provide high impact protection
Manufacturer: Protective Industrial Products, Inc. 253OR40056
 View more versions of this product
Catalog No. 19-036-825
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Weight (Metric) 43g
    Lens Coating Anti-scratch
    Temple Style Adjustable
    Quantity One pair/pk.
    For Use With (Application) Assembly and production work; Light engineering; Cutting/Brazing/Torch work
    Includes Soft pouch and adjustable cord
    Lens Material Polycarbonate
  • Description & Specifications
Related Products
Product Content Feedback