PIP™ Bolle's Safety Solis Protective Eyewear
Light weight hydrated nylon frame front for extensive eye coverage
Manufacturer: Protective Industrial Products, Inc. 253-SS-40062
 View more versions of this product
Catalog No. 19-036-831
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Weight (Metric) 25g
    Frame Color Black
    Lens Coating Anti-scratch; Anti-fog
    Quantity One pair/pk.
    Certifications/Compliance ANSI Z87.1-2010
    Includes Soft pouch and adjustable cord
    Lens Material Polycarbonate
    Lens Tint Clear
  • Description & Specifications
Related Products
Product Content Feedback